Product Details

SNP ID
rs11952641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150221384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGTGCTTTGCTGTGGTCATCA[A/G]ACGCCAAGGGGAGAGAGGCAATGAA
Phenotype
MIM: 114078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMK2A PubMed Links

Gene Details

Gene
CAMK2A
Gene Name
calcium/calmodulin dependent protein kinase II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015981.3 2978 UTR 3 NP_057065.2
NM_171825.2 2978 UTR 3 NP_741960.1
XM_017009898.1 2978 Intron XP_016865387.1

View Full Product Details