Product Details
- SNP ID
-
rs4672766
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:215318046 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATACTTTAAAAAATCAGAATTTTC[A/T]CGTTGAATTCAGGCTCAAAATCTCT
- Phenotype
-
MIM: 601731
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATIC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75285060] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATIC
- Gene Name
- 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
View Full Product Details