Product Details

SNP ID
rs13428084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232621892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCACCCCTCTGTGCCCATGACA[A/G]AGGGCTCCTGGTTGTGTCTGAAGGG
Phenotype
MIM: 611617
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFHD1 PubMed Links

Gene Details

Gene
EFHD1
Gene Name
EF-hand domain family member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243252.1 Intron NP_001230181.1
NM_001308395.1 Intron NP_001295324.1
NM_025202.3 Intron NP_079478.1
XM_011511924.2 Intron XP_011510226.1

View Full Product Details