Product Details
- SNP ID
-
hCV31243272
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:44105743 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATCATATAGGAGAGGCTTGTTTG[C/G]TAAGGATCTTAAAATGTGGTCTACA
- Phenotype
-
MIM: 608210
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EXT2
PubMed Links
Gene Details
- Gene
- EXT2
- Gene Name
- exostosin glycosyltransferase 2
There are no transcripts associated with this gene.
View Full Product Details