Product Details
- SNP ID
-
hCV31247845
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:14462586 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTCAACTTTCAGGGAAAACTGA[C/G]GCTACCAAGAATAAGCTCCTTTAAT
- Phenotype
-
MIM: 600959
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
COPB1
PubMed Links
Gene Details
- Gene
- COPB1
- Gene Name
- coatomer protein complex subunit beta 1
There are no transcripts associated with this gene.
View Full Product Details