Product Details

SNP ID
rs11020996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94985519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTAATGCTCTACAAATTGATCTC[C/T]AGATTGATGCAATCCTACCATCATT
Phenotype
MIM: 609766
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM4D PubMed Links

Gene Details

Gene
KDM4D
Gene Name
lysine demethylase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018039.2 Intron NP_060509.2

View Full Product Details