Product Details

SNP ID
rs12295932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95095669 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATTTATTGATCCCTTACCATGTC[A/C]AAGGTCTTCACATTCTAACTCATCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENDOD1 PubMed Links

Gene Details

Gene
ENDOD1
Gene Name
endonuclease domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015036.2 Intron NP_055851.1

View Full Product Details