Product Details

SNP ID
rs10893426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125778426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACAAAACAGGTCTGGCAGGAGG[C/T]GAAACCCCTAGGATTCTATAATAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PATE2 PubMed Links

Gene Details

Gene
PATE2
Gene Name
prostate and testis expressed 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_212555.2 Intron NP_997720.1
Gene
PATE3
Gene Name
prostate and testis expressed 3
There are no transcripts associated with this gene.

View Full Product Details