Product Details

SNP ID
rs12363776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129904305 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGAGCATTTTTCCTTTTTTTTCC[C/T]TCTAGCTTTATTAAGATATAATTAA
Phenotype
MIM: 164013
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFRKB PubMed Links

Gene Details

Gene
NFRKB
Gene Name
nuclear factor related to kappaB binding protein
There are no transcripts associated with this gene.

Gene
PRDM10
Gene Name
PR domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020228.2 Intron NP_064613.2
NM_199437.1 Intron NP_955469.1
NM_199438.1 Intron NP_955470.1
NM_199439.1 Intron NP_955471.1
XM_011542912.2 Intron XP_011541214.1
XM_017018023.1 Intron XP_016873512.1

View Full Product Details