Product Details

SNP ID
rs11248341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122934269 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACACATATTTATTTAAACACCACC[A/G]TAATCAAAAGGCAGAACCATTCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf88 PubMed Links

Gene Details

Gene
C10orf88
Gene Name
chromosome 10 open reading frame 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024942.3 Intron NP_079218.2

View Full Product Details