Product Details
- SNP ID
-
rs10885325
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:112284028 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACTTTTTTTTTAATTAGAGAATGC[C/T]TGCTCTATTTCACTCTCTTACTTAC
- Phenotype
-
MIM: 602653
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TECTB
PubMed Links
Gene Details
- Gene
- TECTB
- Gene Name
- tectorin beta
View Full Product Details