Product Details

SNP ID
rs12253626
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104032161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATTGACAGACTCCAGCTTTCAC[A/C]CTCTGGAGACCTTGGACCTAAGTGC
Phenotype
MIM: 113811 MIM: 616563
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
COL17A1 PubMed Links

Gene Details

Gene
COL17A1
Gene Name
collagen type XVII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000494.3 4737 UTR 3 NP_000485.3
Gene
SLK
Gene Name
STE20 like kinase
There are no transcripts associated with this gene.

View Full Product Details