Product Details

SNP ID
rs11191887
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103975136 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTTAAGAGAACCGGTCTTCTC[A/C]CATGGCTTCAATTACCATCTAAGTG
Phenotype
MIM: 616563
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
SLK PubMed Links
Additional Information
For this assay, SNP(s) [rs76416280] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SLK
Gene Name
STE20 like kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304743.1 Intron NP_001291672.1
NM_014720.3 Intron NP_055535.2
XM_011540401.2 Intron XP_011538703.1

View Full Product Details