Product Details

SNP ID
rs12766020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97001652 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGCTGGAGGCCCCGGGGGCAAT[A/G]ACATTATGAAAAGGATTCATGCTGG
Phenotype
MIM: 603742
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLIT1 PubMed Links

Gene Details

Gene
SLIT1
Gene Name
slit guidance ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003061.2 Intron NP_003052.2

View Full Product Details