Product Details
- SNP ID
-
hCV31282862
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:129626621 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAATGAATGCTGAATTTAATGTT[A/C]CTGGGTGACTTAAAGGAGCATTTAA
- Phenotype
-
MIM: 600879
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NRF1
PubMed Links
Gene Details
- Gene
- NRF1
- Gene Name
- nuclear respiratory factor 1
There are no transcripts associated with this gene.
View Full Product Details