Product Details

SNP ID
rs12378288
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33441433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATGTACATCCAGAGCTCCAAAC[C/G]TAAGCTGTGGCCCCCTCCTCCCAGC
Phenotype
MIM: 600170
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
AQP3 PubMed Links

Gene Details

Gene
AQP3
Gene Name
aquaporin 3 (Gill blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318144.1 1592 UTR 3 NP_001305073.1
NM_004925.4 1592 UTR 3 NP_004916.1

View Full Product Details