Product Details

SNP ID
rs11015594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27188369 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAAAAGTTTGGGAATCAAACAC[A/G]AATTGTTGAAATCACTGCTCTGCCA
Phenotype
MIM: 616618 MIM: 608221
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACBD5 PubMed Links

Gene Details

Gene
ACBD5
Gene Name
acyl-CoA binding domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042473.3 Intron NP_001035938.1
NM_001271512.3 Intron NP_001258441.1
NM_001301251.1 Intron NP_001288180.1
NM_001301252.1 Intron NP_001288181.1
NM_001301253.1 Intron NP_001288182.1
NM_001301254.1 Intron NP_001288183.1
NM_145698.4 Intron NP_663736.2
XM_006717528.3 Intron XP_006717591.3
XM_006717530.3 Intron XP_006717593.1
XM_006717531.2 Intron XP_006717594.1
XM_006717535.2 Intron XP_006717598.1
XM_017016884.1 Intron XP_016872373.1
XM_017016885.1 Intron XP_016872374.1
XM_017016886.1 Intron XP_016872375.1
XM_017016887.1 Intron XP_016872376.1
XM_017016888.1 Intron XP_016872377.1
XM_017016889.1 Intron XP_016872378.1
XM_017016890.1 Intron XP_016872379.1
XM_017016891.1 Intron XP_016872380.1
XM_017016892.1 Intron XP_016872381.1
XM_017016893.1 Intron XP_016872382.1
XM_017016894.1 Intron XP_016872383.1
XM_017016895.1 Intron XP_016872384.1
XM_017016896.1 Intron XP_016872385.1
XM_017016897.1 Intron XP_016872386.1
XM_017016898.1 Intron XP_016872387.1
XM_017016899.1 Intron XP_016872388.1
XM_017016900.1 Intron XP_016872389.1
XM_017016901.1 Intron XP_016872390.1
XM_017016902.1 Intron XP_016872391.1
XM_017016903.1 Intron XP_016872392.1
XM_017016904.1 Intron XP_016872393.1
XM_017016905.1 Intron XP_016872394.1
XM_017016906.1 Intron XP_016872395.1
XM_017016907.1 Intron XP_016872396.1
XM_017016908.1 Intron XP_016872397.1
Gene
MASTL
Gene Name
microtubule associated serine/threonine kinase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172303.2 Intron NP_001165774.1
NM_001172304.2 Intron NP_001165775.1
NM_001320756.1 Intron NP_001307685.1
NM_001320757.1 Intron NP_001307686.1
NM_032844.4 Intron NP_116233.2
XM_005252631.3 Intron XP_005252688.1
XM_005252632.3 Intron XP_005252689.1
XM_006717519.3 Intron XP_006717582.1
XM_006717520.3 Intron XP_006717583.1
XM_011519751.2 Intron XP_011518053.1
XM_017016852.1 Intron XP_016872341.1
XM_017016853.1 Intron XP_016872342.1
XM_017016854.1 Intron XP_016872343.1
XM_017016855.1 Intron XP_016872344.1
XM_017016856.1 Intron XP_016872345.1

View Full Product Details