Product Details
- SNP ID
-
rs12852974
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:15685881 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTTGAAGAGCCCAGGGACATACA[A/G]GGAGGCTAGAGAAATAGGTGGTGCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CA5BP1
PubMed Links
Gene Details
- Gene
- CA5BP1
- Gene Name
- carbonic anhydrase 5B pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details