Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_144974.3 | 248 | Intron | NP_659411.2 | ||
XM_017020397.1 | 248 | Intron | XP_016875886.1 | ||
XM_017020398.1 | 248 | Intron | XP_016875887.1 | ||
XM_017020399.1 | 248 | Intron | XP_016875888.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001128303.1 | 248 | Intron | NP_001121775.1 | ||
NM_153218.2 | 248 | UTR 5 | NP_694950.2 | ||
XM_005266261.3 | 248 | Intron | XP_005266318.1 | ||
XM_006719766.3 | 248 | Intron | XP_006719829.1 | ||
XM_006719767.3 | 248 | UTR 5 | XP_006719830.1 | ||
XM_011534933.2 | 248 | UTR 5 | XP_011533235.1 | ||
XM_011534934.2 | 248 | UTR 5 | XP_011533236.1 | ||
XM_011534935.2 | 248 | UTR 5 | XP_011533237.1 | ||
XM_017020394.1 | 248 | UTR 5 | XP_016875883.1 |