Product Details

SNP ID
hCV31317217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57389001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCACCTCAGCCTCAGCCATAG[G/T]CCACTCACCCAAGCTTGACTAAACG
Phenotype
MIM: 605601 MIM: 606814
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRG2 PubMed Links

Gene Details

Gene
PRG2
Gene Name
proteoglycan 2, pro eosinophil major basic protein
There are no transcripts associated with this gene.

Gene
PRG3
Gene Name
proteoglycan 3, pro eosinophil major basic protein 2
There are no transcripts associated with this gene.

View Full Product Details