Product Details

SNP ID
rs11229025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57408653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAAGATTTGCTGCTTTAAAAAT[A/T]AAAAATGAGGCAAATAGAGCTGTGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC43A3 PubMed Links

Gene Details

Gene
SLC43A3
Gene Name
solute carrier family 43 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278201.1 Intron NP_001265130.1
NM_001278206.1 Intron NP_001265135.1
NM_014096.3 Intron NP_054815.2
NM_017611.2 Intron NP_060081.1
NM_199329.2 Intron NP_955361.1

View Full Product Details