Product Details

SNP ID
rs12279957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58402633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCATGGAGTGACTGGAGCTGGG[C/T]TGTAAGTACATGAAGATAATAGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5B3 PubMed Links

Gene Details

Gene
OR5B3
Gene Name
olfactory receptor family 5 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005469.1 777 Silent Mutation CAA,CAG Q259Q NP_001005469.1

View Full Product Details