Product Details

SNP ID
rs12223473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59123019 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATATAATATTTTTTACTTATGTT[G/T]TTCATTTCTGTTGCCATCAGATTCT
Phenotype
MIM: 615584
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM111B PubMed Links

Gene Details

Gene
FAM111B
Gene Name
family with sequence similarity 111 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142703.1 Intron NP_001136175.1
NM_001142704.1 Intron NP_001136176.1
NM_198947.3 Intron NP_945185.1

View Full Product Details