Product Details

SNP ID
rs12285568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67614002 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAACCCCACCCTCTTCACCCCCT[A/G]TTTCCCTGGGGACCAGATGCTGAGT
Phenotype
MIM: 161015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf72 PubMed Links

Gene Details

Gene
C11orf72
Gene Name
chromosome 11 open reading frame 72
There are no transcripts associated with this gene.

Gene
DOC2GP
Gene Name
double C2 domain gamma pseudogene
There are no transcripts associated with this gene.

Gene
NDUFV1
Gene Name
NADH:ubiquinone oxidoreductase core subunit V1
There are no transcripts associated with this gene.

Gene
NUDT8
Gene Name
nudix hydrolase 8
There are no transcripts associated with this gene.

View Full Product Details