Product Details

SNP ID
rs11233392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82976775 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAAGAGCCGTCTATCAGCCTTG[C/G]AAGGGTGAAAGTTATTTTGCAGGTG
Phenotype
MIM: 605693
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB30 PubMed Links

Gene Details

Gene
RAB30
Gene Name
RAB30, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286059.1 6141 UTR 3 NP_001272988.1
NM_001286060.1 6141 UTR 3 NP_001272989.1
NM_001286061.1 6141 UTR 3 NP_001272990.1
NM_014488.4 6141 UTR 3 NP_055303.2

View Full Product Details