Product Details

SNP ID
rs11622442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20143815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCTCAAGATGCTCAACTTCTGG[G/T]CTTTGTGCTAGTCTTAATTTTCTAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4N5 PubMed Links

Gene Details

Gene
OR4N5
Gene Name
olfactory receptor family 4 subfamily N member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004724.1 80 Missense Mutation GGC,GTC G27V NP_001004724.1

View Full Product Details