Product Details
- SNP ID
-
rs13051540
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:44456480 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCTCCCTCCCTCTCCGCTCCCTC[C/T]TGCGTCTTCCCTTTCAGAGCTGCCA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112589449] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC3
- Gene Name
- leucine rich repeat containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030891.4 |
|
Intron |
|
|
NP_112153.1 |
- Gene
- LRRC3-AS1
- Gene Name
- LRRC3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details