Product Details

SNP ID
rs13202369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39054922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCACCTCCTCAGCTGTGGCTAAT[A/G]TCATGGACAACACCTTTGCAGCTGT
Phenotype
MIM: 138032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLP1R PubMed Links

Gene Details

Gene
GLP1R
Gene Name
glucagon like peptide 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002062.4 Intron NP_002053.3
XM_017010750.1 Intron XP_016866239.1
XM_017010751.1 Intron XP_016866240.1

View Full Product Details