Product Details

SNP ID
rs7096663
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59793743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTTGCAGTGAGCAGACATCATGC[C/T]GCTGAACTCCAGCCTGGATGACAGA
Phenotype
MIM: 601985
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC6 PubMed Links

Gene Details

Gene
CCDC6
Gene Name
coiled-coil domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005436.4 Intron NP_005427.2

View Full Product Details