Product Details
- SNP ID
-
rs10761781
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:63524843 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAAGTCATTATAATAAATACCACA[C/T]GAAACCGTTGTAAAGAAAATTAGCC
- Phenotype
-
MIM: 604503
MIM: 609348
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
JMJD1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115738314] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- JMJD1C
- Gene Name
- jumonji domain containing 1C
There are no transcripts associated with this gene.
- Gene
- REEP3
- Gene Name
- receptor accessory protein 3
View Full Product Details