Product Details

SNP ID
rs10997817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:67804053 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCATCTAACTATCTTCTTTTGAAT[A/G]CAAGTGTTCTAGGGTGTTCAGGAAC
Phenotype
MIM: 606060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC12 PubMed Links

Gene Details

Gene
DNAJC12
Gene Name
DnaJ heat shock protein family (Hsp40) member C12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021800.2 Intron NP_068572.1
NM_201262.1 Intron NP_957714.1
XM_017016431.1 Intron XP_016871920.1
XM_017016432.1 Intron XP_016871921.1

View Full Product Details