Product Details
- SNP ID
-
hCV31353928
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:83640457 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGGTTGGGTTCTCTCGTGTATAT[A/C]CAAATAGGAATAATAAGAATTTTGA
- Phenotype
-
MIM: 115435
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HAPLN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78259722] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAPLN1
- Gene Name
- hyaluronan and proteoglycan link protein 1
There are no transcripts associated with this gene.
View Full Product Details