Product Details

SNP ID
hCV31361273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24704921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATTTTATTTAGGTTTTCTTTTT[C/T]TTTTTTTCTTTTTTTTTCAAATTCC
Phenotype
MIM: 615652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT13 PubMed Links

Gene Details

Gene
ACOT13
Gene Name
acyl-CoA thioesterase 13
There are no transcripts associated with this gene.

Gene
C6orf62
Gene Name
chromosome 6 open reading frame 62
There are no transcripts associated with this gene.

View Full Product Details