Product Details

SNP ID
rs6559753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83989896 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTGAAGAGAGATCTGCATTCCAG[C/T]GTTTATTACAGTACTATTTACAGTT
Phenotype
MIM: 600712 MIM: 610404
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPK PubMed Links

Gene Details

Gene
HNRNPK
Gene Name
heterogeneous nuclear ribonucleoprotein K
There are no transcripts associated with this gene.

Gene
RMI1
Gene Name
RecQ mediated genome instability 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024945.2 Intron NP_079221.2
XM_005252211.2 Intron XP_005252268.1
XM_005252213.2 Intron XP_005252270.1
XM_011519034.2 Intron XP_011517336.1
XM_017015140.1 Intron XP_016870629.1

View Full Product Details