Product Details
- SNP ID
-
rs13243262
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:80743783 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGACAGAATGTCTCCTCATATTG[G/T]TTTCTTACTTTCTAGACATAACAAG
- Phenotype
-
MIM: 602645
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SEMA3C
PubMed Links
Gene Details
- Gene
- SEMA3C
- Gene Name
- semaphorin 3C
View Full Product Details