Product Details

SNP ID
rs11047791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25058131 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATCTAGGAGTAGAACTGTTAGG[A/T]CATAGCATAGCTGTGACTGTCACTT
Phenotype
MIM: 602003
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LRMP PubMed Links

Gene Details

Gene
LRMP
Gene Name
lymphoid restricted membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204126.1 Intron NP_001191055.1
NM_001204127.1 Intron NP_001191056.1
NM_006152.3 Intron NP_006143.2
XM_005253370.3 Intron XP_005253427.1
XM_005253372.2 Intron XP_005253429.1
XM_005253374.3 Intron XP_005253431.1
XM_006719076.2 Intron XP_006719139.1
XM_011520667.1 Intron XP_011518969.1
XM_011520668.2 Intron XP_011518970.1
XM_011520669.2 Intron XP_011518971.1
XM_011520670.1 Intron XP_011518972.1
XM_017019300.1 Intron XP_016874789.1
XM_017019301.1 Intron XP_016874790.1
XM_017019302.1 Intron XP_016874791.1

View Full Product Details