Product Details

SNP ID
rs11047887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25195738 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGTCCTTTTTCCTAAGAGCGCCCT[A/C]AACAAGAATTGAAAATGAAGCAATT
Phenotype
MIM: 616906 MIM: 190070
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CASC1 PubMed Links

Gene Details

Gene
CASC1
Gene Name
cancer susceptibility candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082972.2 Intron NP_001076441.1
NM_001082973.2 Intron NP_001076442.1
NM_001204101.2 Intron NP_001191030.1
NM_001204102.2 Intron NP_001191031.1
NM_001319977.1 Intron NP_001306906.1
NM_001319978.1 Intron NP_001306907.1
NM_018272.4 Intron NP_060742.3
XM_005253406.1 Intron XP_005253463.1
XM_005253408.1 Intron XP_005253465.1
XM_005253409.1 Intron XP_005253466.1
XM_006719101.1 Intron XP_006719164.1
XM_011520723.1 Intron XP_011519025.1
XM_011520726.1 Intron XP_011519028.1
XM_011520727.1 Intron XP_011519029.1
XM_017019560.1 Intron XP_016875049.1
XM_017019561.1 Intron XP_016875050.1
XM_017019562.1 Intron XP_016875051.1
XM_017019563.1 Intron XP_016875052.1
XM_017019564.1 Intron XP_016875053.1
XM_017019565.1 Intron XP_016875054.1
XM_017019566.1 Intron XP_016875055.1
XM_017019567.1 Intron XP_016875056.1
XM_017019568.1 Intron XP_016875057.1
Gene
KRAS
Gene Name
KRAS proto-oncogene, GTPase
There are no transcripts associated with this gene.

Gene
LYRM5
Gene Name
LYR motif containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001660.2 Intron NP_001001660.2
XM_005253319.4 Intron XP_005253376.1
XM_005253320.4 Intron XP_005253377.1
XM_017018850.1 Intron XP_016874339.1

View Full Product Details