Product Details
- SNP ID
-
rs11048819
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:27023192 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGGCCCAGAGTATCCTTCCATCT[C/G]TTGCCTCTTGGCCATCCCTCAGAAT
- Phenotype
-
MIM: 603800
MIM: 605181
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MED21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140399543] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MED21
- Gene Name
- mediator complex subunit 21
- Gene
- TM7SF3
- Gene Name
- transmembrane 7 superfamily member 3
There are no transcripts associated with this gene.
View Full Product Details