Product Details

SNP ID
rs12231079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27028177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGATTTTTAGAATTTCTTTACTG[A/G]AAAACATATCACAATGATTGCAAAG
Phenotype
MIM: 603800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED21 PubMed Links

Gene Details

Gene
MED21
Gene Name
mediator complex subunit 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271811.1 Intron NP_001258740.1
NM_004264.4 Intron NP_004255.2

View Full Product Details