Product Details
- SNP ID
-
rs12321184
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:29433031 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAAAAGGGGAGACAGTGAGTATAA[A/G]TAACTTCCATGGAGTCTTGCTAAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OVCH1
PubMed Links
Gene Details
- Gene
- OVCH1
- Gene Name
- ovochymase 1
- Gene
- OVCH1-AS1
- Gene Name
- OVCH1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details