Product Details

SNP ID
rs12317348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:30928794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCCTTTCCTGATCCCCCATCT[A/G]AAGTAGCCACCTCCCAGCCATGATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN11 PubMed Links

Gene Details

Gene
TSPAN11
Gene Name
tetraspanin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080509.2 Intron NP_001073978.1
XM_011520679.2 Intron XP_011518981.1
XM_011520680.2 Intron XP_011518982.1

View Full Product Details