Product Details
- SNP ID
-
rs11049127
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:27769141 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAGAAAGAATAAGACAAAAATTAG[C/T]TAATAATACATTGCTAATACATTAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MANSC4
PubMed Links
Gene Details
- Gene
- MANSC4
- Gene Name
- MANSC domain containing 4
View Full Product Details