Product Details

SNP ID
rs12566231
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:100900172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGGTTTTTTCTAGTATCTCAAT[A/G]TAGTTTCTGTGTTTTTATGTTCTTA
Phenotype
MIM: 602411 MIM: 611149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXTL2 PubMed Links

Gene Details

Gene
EXTL2
Gene Name
exostosin like glycosyltransferase 2
There are no transcripts associated with this gene.

Gene
SLC30A7
Gene Name
solute carrier family 30 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144884.1 Intron NP_001138356.1
NM_133496.4 Intron NP_598003.2
XM_011540779.2 Intron XP_011539081.1
XM_017000400.1 Intron XP_016855889.1
XM_017000401.1 Intron XP_016855890.1

View Full Product Details