Product Details
- SNP ID
-
rs12563154
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
55
- Location
-
Chr.1:95096631 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAAGCCCCTATTACACAACTCTTA[C/T]ATTATATAGTGTTTCATGAAGTATC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM56
PubMed Links
Gene Details
- Gene
- TMEM56
- Gene Name
- transmembrane protein 56
View Full Product Details