Product Details

SNP ID
rs11576568
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:228086542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAAAAGAAAAAAAAGAA[A/T]CAGCATGAAATTCCTTGTTGATGGG
Phenotype
MIM: 103180
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARF1 PubMed Links

Gene Details

Gene
ARF1
Gene Name
ADP ribosylation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024226.1 Intron NP_001019397.1
NM_001024227.1 Intron NP_001019398.1
NM_001024228.1 Intron NP_001019399.1
NM_001658.3 Intron NP_001649.1
XM_017001233.1 Intron XP_016856722.1
Gene
LOC100506571
Gene Name
uncharacterized LOC100506571
There are no transcripts associated with this gene.

View Full Product Details