Product Details

SNP ID
rs10916265
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:228089347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAAGCGAGGATTTCCTGCAAGG[A/G]CGGTGGCCTCCCAGGGTCTGTGGGG
Phenotype
MIM: 103180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARF1 PubMed Links

Gene Details

Gene
ARF1
Gene Name
ADP ribosylation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024226.1 Intron NP_001019397.1
NM_001024227.1 Intron NP_001019398.1
NM_001024228.1 Intron NP_001019399.1
NM_001658.3 Intron NP_001649.1
XM_017001233.1 Intron XP_016856722.1
Gene
LOC100506571
Gene Name
uncharacterized LOC100506571
There are no transcripts associated with this gene.

Gene
MIR3620
Gene Name
microRNA 3620
There are no transcripts associated with this gene.

View Full Product Details