Product Details

SNP ID
rs12841236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:86148957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTGCACACTGTGTACACCAAG[C/T]TGAAGAGACTGGATATATCCCCCGT
Phenotype
MIM: 300608
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DACH2 PubMed Links

Gene Details

Gene
DACH2
Gene Name
dachshund family transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139514.1 506 Silent Mutation CTG,TTG L113L NP_001132986.1
NM_001139515.1 506 Intron NP_001132987.1
NM_053281.3 506 Silent Mutation CTG,TTG L113L NP_444511.1
XM_011530846.2 506 Silent Mutation CTG,TTG L113L XP_011529148.1
XM_011530847.2 506 Silent Mutation CTG,TTG L113L XP_011529149.1
XM_011530848.2 506 Silent Mutation CTG,TTG L113L XP_011529150.1
XM_017029254.1 506 Silent Mutation CTG,TTG L113L XP_016884743.1
XM_017029255.1 506 Silent Mutation CTG,TTG L113L XP_016884744.1
XM_017029256.1 506 Silent Mutation CTG,TTG L113L XP_016884745.1

View Full Product Details