Product Details

SNP ID
rs13339741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12394845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGCCTTCTCACCAAGAAAGTG[C/T]AGCATTCCAAAAACAAATAAAGGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF799 PubMed Links

Gene Details

Gene
ZNF799
Gene Name
zinc finger protein 799
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080821.2 Intron NP_001074290.1
NM_001322497.1 Intron NP_001309426.1
NM_001322498.1 Intron NP_001309427.1
XM_017027452.1 Intron XP_016882941.1

View Full Product Details