Product Details

SNP ID
rs12841355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:109537720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGTTCAGGTGGTGTCATTTTCA[A/T]TGCCGTGAATGTTTGTAAAATATTG
Phenotype
MIM: 300320
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NXT2 PubMed Links

Gene Details

Gene
NXT2
Gene Name
nuclear transport factor 2 like export factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242617.1 Intron NP_001229546.1
NM_001242618.1 Intron NP_001229547.1
NM_018698.4 Intron NP_061168.2
XM_011530989.2 Intron XP_011529291.1
XM_017029648.1 Intron XP_016885137.1

View Full Product Details