Product Details

SNP ID
rs11579196
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156808303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGCTGGAGAGTTAGCAATCAGG[A/C]CAGTGAACATGGGACCGAGCTGTTT
Phenotype
MIM: 191315 MIM: 179755 MIM: 604514
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NTRK1 PubMed Links

Gene Details

Gene
NTRK1
Gene Name
neurotrophic receptor tyrosine kinase 1
There are no transcripts associated with this gene.

Gene
PRCC
Gene Name
papillary renal cell carcinoma (translocation-associated)
There are no transcripts associated with this gene.

Gene
SH2D2A
Gene Name
SH2 domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161441.1 Intron NP_001154913.1
NM_001161442.1 Intron NP_001154914.1
NM_001161443.1 Intron NP_001154915.1
NM_001161444.1 Intron NP_001154916.1
NM_003975.3 Intron NP_003966.2
XM_006711615.2 Intron XP_006711678.1
XM_017002762.1 Intron XP_016858251.1
XM_017002763.1 Intron XP_016858252.1
XM_017002764.1 Intron XP_016858253.1
XM_017002765.1 Intron XP_016858254.1
XM_017002766.1 Intron XP_016858255.1
XM_017002767.1 Intron XP_016858256.1

View Full Product Details